Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PPM1G protein.

Manufacturer:  Abnova Corporation H00005496B01

Catalog No. 89-006-002


Add to cart

Description

Description

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq

Sequence: MGAYLSQPNTVKCSGDGVGAPRLPLPYGFSAMQGWRVSMEDAHNCIPELDSETAMFSVYDGHGGEEVALYCAKYLPDIIKDQKAYKEGKLQKALEDAFLAIDAKLTTEEVIKELAQIAGRPTEDEDEKEKVADEDDVDNEEAALLHEEATMTIEELLTRYGQNCHKGPPHSKSGGGTGEEPGSQGLNGEAGPEDSTRETPSQENGPTAKAYTGFSSNSERGTEAGQVGEPGIPTGEAGPSCSSASDKLPRVAKSKFFEDSEDESDEAEEEEEDSEECSEEEDGYSSEEAENEEDEDDTEEAEEDDEEEEEEMMVPGMEGKEEPGSDSGTTAVVALIRGKQLIVANAGDSRCVVSEAGKALDMSYDHKPEDEVELARIKNAGGKVTMDGRVNGGLNLSRAIGDHFYKRNKNLPPEEQMISALPDIKVLTLTDDHEFMVIACDGIWNVMSSQEVVDFIQSKISQRDENGELRLLSSIVEELLDQCLAPDTSGDGTGCDNMTCIIICFKPRNTAELQPESGKRKLEEVLSTEGAEENGNSDKKKKAKRD
Specifications

Specifications

protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform
Unconjugated
No additive
NM_002707
MGC1675|MGC2870|PP2CG|PP2CGAMMA|PPP2CG
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human PPM1G protein.
PPM1G
Serum
PPM1G
PPM1G (NP_002698, 1 a.a. ~ 546 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5496
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit