Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1J Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527891

Catalog No. PIPA527891


Add to cart

Description

Description

PA5-27891 targets PPM1J in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27891 immunogen is recombit fragment corresponding to a region within amino acids 133 and 442 of Human PPM1J. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes the serine/threonine protein phosphatase. The mouse homolog of this gene apparently belongs to the protein phosphatase 2C family of genes. The exact function of this gene is not yet known.
Specifications

Specifications

PPM1J
Polyclonal
Unconjugated
PPM1J
Liquid
PPM1J
Recombit fragment corresponding to a region within amino acids 133 and 442 of Human PPM1J
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q5JR12
PP2CZ, PP2Czeta, PPP2CZ, PP2C-zeta, protein phosphatase 1J, protein phosphatase 1J (PP2C domain containing), protein phosphatase 2C isoform zeta, protein phosphatase 2C zeta, protein phosphatase 2a, catalytic subunit, zeta isoform
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
333926
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit