Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524480

Catalog No. PIPA524480


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK; see MIM 601158) signaling cascades that respond to extracellular stimuli.
Specifications

Specifications

PPM1L
Polyclonal
PBS with 0.09% sodium azide
Q5SGD2, Q8BHN0
PP2C-epsilon, PP2CE, PPM1-LIKE, PP2C epsilon, Protein phosphatase 2C epsilon isoform, protein phosphatase 1L, protein phosphatase 2C isoform epsilon
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
151742, 242083
Immunohistochemistry, Western Blot
Unconjugated
PPM1L
Liquid
PPM1L
KLH conjugated synthetic peptide between 215-244 amino acids from the C-terminal region of human PPM1L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit