Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524170

Catalog No. PIPA524170


Add to cart

Description

Description

This antibody is predicted to react with rabbit based on sequence homology.

The glycogen-associated form of protein phosphatase-1 (PP1) derived from skeletal muscle is a heterodimer composed of a 37-kD catalytic subunit and a 124-kD targeting and regulatory subunit. This gene encodes the regulatory subunit which binds to muscle glycogen with high affinity, thereby enhancing dephosphorylation of glycogen-bound substrates for PP1 such as glycogen synthase and glycogen phosphorylase kinase.
Specifications

Specifications

PPP1R3A
Polyclonal
PBS with 0.09% sodium azide
Q16821
GM, PP1G, PPP1R3, RG1, glycogen and sarcoplasmic reticulum binding subunit, skeletal muscle, glycogen-associated regulatory subunit of protein phosphatase-1, protein phosphatase 1 glycogen- associated regulatory subunit, protein phosphatase 1 glycogen-associated regulatory sub
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5506
Flow Cytometry, Western Blot
Unconjugated
PPP1R3A
Liquid
PPP1R3A
KLH conjugated synthetic peptide between 754-782 amino acids from the central region of human PPP1R3A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit