Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PPP2CB, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132069

Catalog No. NBP132069


Add to cart

Description

Description

PPP2CB Polyclonal antibody specifically detects PPP2CB in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PPP2CB
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
PPP2CB
Recombinant protein encompassing a sequence within the center region of human PP2A beta. The exact sequence is proprietary.
36 kDa
100 ul
Wnt Signaling Pathway
Primary
5516
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
EC 3.1.3.16, PP2Abeta, PP2A-beta, PP2CB, protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isozyme, protein phosphatase 2A catalytic subunit, beta isoform, protein phosphatase type 2A catalytic subunit, serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (100%), Xenopus tropicalis (100%), and Rhesus Monkey (100%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only