Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PPP2R2B, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133043

Catalog No. NBP133043


Add to cart

Description

Description

PPP2R2B Polyclonal antibody specifically detects PPP2R2B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PPP2R2B
0.7mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Q00005
B55BETA, FLJ95686, MGC24888, PP2A subunit B isoform B55-beta, PP2A subunit B isoform beta, PP2A subunit B isoform PR55-beta, PP2A subunit B isoform R2-beta, PP2A, subunit B, B-beta isoform, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55BETA, PR55-BETA, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform, protein phosphatase 2, regulatory subunit B, beta, SCA12, serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, betaisoform, serine/threonine protein phosphatase 2A, neuronal isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B betaisoform, spinocerebellar ataxia 12
Rabbit
IgG
0.1 ml
Stem Cell Markers
Primary
5521
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
PPP2R2B
Recombinant protein encompassing a sequence within the center region of human PPP2R2B. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human and Mouse. Predicted cross-reactivity based on sequence identity: Pig (90%), Rabbit (100%), Rat (99%), Xenopus laevis (94%), Bovine (100%), Xenopus tropicalis (94%).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only