Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PPP2R3A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 46340002

Catalog No. 46-340-002


Add to cart

Description

Description

PPP2R3A Polyclonal antibody specifically detects PPP2R3A in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PPP2R3A
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
PPP2R3A
This antibody is specific for the N Terminus Region of the target protein.
Immunogen affinity purified
RUO
Primary
5523
Human
ELISA, Western Blot
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
Q06190
PP2A subunit B isoform R3 isoform, PP2A subunit B isoforms B72/B130, PP2A subunit B isoforms B'-PR72/PR130, protein phosphatase 2 (formerly 2A), regulatory subunit B' (PR 72), alphaisoform and (PR 130), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha, serine/threonine-protein phosphatase 2A regulatory subunit B' subunit alpha, subunit B, R3 isoform
Rabbit
IgG
0.05 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human PPP2R3A
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only