Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein phosphatase 2, regulatory subunit B', epsilon isoform, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PPP2R5E protein.

Manufacturer:  Abnova Corporation H00005529B01

Catalog No. 89-006-014


Add to cart

Description

Description

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily. [provided by RefSeq

Sequence: MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQPSIAKKYIDQKFVLQLLELFDSEDPRERDYLKTVLHRIYGKFLGLRAFIRKQINNIFLRFVYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLVKVLIPLHTVRSLSLFHAQLAYCIVQFLEKDPSLTEPVIRGLMKFWPKTCSQKEVMFLGELEEILDVIEPSQFVKIQEPLFKQIAKCVSSPHFQVAERALYYWNNEYIMSLIEENSNVILPIMFSSLYRISKEHWNPAIVALVYNVLKAFMEMNSTMFDELTATYKSDRQREKKKEKEREELWKKLEDLELKRGLRRDGIIPT
Specifications

Specifications

protein phosphatase 2, regulatory subunit B', epsilon isoform
Unconjugated
No additive
NM_006246.2
PPP2R5E
PPP2R5E (NP_006237.1, 1 a.a. ~ 467 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human PPP2R5E protein.
PPP2R5E
Serum
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5529
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit