Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PPP3CA.

Manufacturer:  Abnova Corporation H00005530A01

Catalog No. 89-004-255


Add to cart

Description

Description

Sequence: MSEPKAIDPKLSTTDRVVKAVPFPPSHRLTAKEVFDNDGKPRVDILKAHLMKEGRLEESVALRIITEGASILRQEKNLLDIDAP
Specifications

Specifications

protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform
Unconjugated
50% glycerol
NM_000944
CALN|CALNA|CALNA1|CCN1|CNA1|PPP2B
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5530
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant PPP3CA.
PPP3CA
Serum
PPP3CA
PPP3CA (NP_000935, 1 a.a. ~ 84 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit