Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP3R2, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a partial recombinant PPP3R2.
Manufacturer: Abnova Corporation H00005535A01
Description
Sequence: MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISpecifications
PPP3R2 | |
Unconjugated | |
50% glycerol | |
NM_147180 | |
PPP3RL | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Polyclonal | |
Human |
ELISA, Western Blot | |
Mouse polyclonal antibody raised against a partial recombinant PPP3R2. | |
PPP3R2 | |
Liquid | |
PPP3R2 | |
PPP3R2 (NP_671709, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
5535 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title