Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP3R2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA526390
Description
PPP3R2 is regulatory subunit of calcineurin, a calcium-dependent, calmodulin stimulated protein phosphatase. This protein confers calcium sensitivity.Specifications
PPP3R2 | |
Polyclonal | |
Liquid | |
PPP3R2 | |
PPP3RL, CBLP, CNBII, calcineurin B, type II (19kDa), calcineurin B-like protein, calcineurin BII, calcineurin subunit B type 2, protein phosphatase 2B regulatory subunit 2, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform (calcineurin B, ty | |
PPP3R2 | |
KLH conjugated synthetic peptide between 3-29 amino acids from the N-terminal region of human PPP3R2 | |
Polyclonal | |
Antigen affinity chromatography | |
RUO | |
Human,Mouse |
Flow Cytometry,Immunohistochemistry,Western Blot | |
Unconjugated | |
PBS with 0.09% sodium azide | |
Q96LZ3, Q63811 | |
19059, 5535 | |
Rabbit | |
IgG | |
Primary | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title