Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PRAK, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP141082

Catalog No. NBP141082


Add to cart

Description

Description

PRAK Polyclonal antibody specifically detects PRAK in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

PRAK
1.0 mg/mL
Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin, Western Blot (Negative)
Affinity Purified
MAPKAPK5
The immunogen for this product maps to a region between residue 250 and 300 of human mitogen-activated protein kinase-activated protein kinase 5 using the numbering given in entry NP_620777.1 (GeneID 8550).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
EC 2.7.11, MAP kinase-activated protein kinase 5, MAPK-activated protein kinase 5, MAPKAP kinase 5, MAPKAPK-5, mitogen-activated protein kinase-activated protein kinase 5, p38-regulated/activated protein kinase, PRAKEC 2.7.11.1
Rabbit
IgG
100 ul
Protein Kinase
Primary
8550
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only