Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAME Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523157

Catalog No. PIPA523157


Add to cart

Description

Description

PRAME is a 509 amino acid protein belonging to the PRAME family consisting of 7 LRR (leucine-rich) repeats. It is a tumor antigen expressed in human melanomas and is recognized by cytolytic T lymphocytes. It ft.s overexpression frequently observed in human cancers confers growth or survival advantages by antagonizing retinoic acid receptor (RAR) signaling. PRAME serves as a candidate marker for monitoring childhood acute leukemia. It is expressed in testis and acute leukemias.
Specifications

Specifications

PRAME
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
P78395
CT130, MAPE, OIP4, OIP-4, Opa-interacting protein OIP4, cancer/testis antigen 130, melanoma antigen preferentially expressed in tumors, opa-interacting protein 4, preferentially expressed antigen of melanoma
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
PRAME
Liquid
PRAME
Synthetic peptide (492 CGDRTFYDPEPIL 502) of PRAME protein.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23532
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit