Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDX3, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127775

Catalog No. PA127775

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in redox regulation of the cell. Protects radical-sensitive enzymes from oxidative damage by a radical-generating system. Acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol. This protein is a protein with an

PA1-27775 detects PRDX3 from human, mouse, and rat samples.PA1-27775 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Thioredoxin-dependent peroxide reductase. Mitochondrial.
Specifications

Specifications

PRDX3
Polyclonal
PBS with no preservative
P30048, P20108, Q9Z0V6
AOP-1, AOP1, MER5, MGC104387, MGC24293, PRO1748, SP-22
Chicken
IgY
50μg
Store at -20°C
Primary
10935
Western Blot
Unconjugated
PRDX3
Purified
PRDX3
Synthetic peptide corresponding to human Thioredoxin-dependent peroxide reductase. Mitochondrial.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit