Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PREB Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1610

Catalog No. PA1610


Add to cart

Description

Description

PA1-610 detects recombit mouse PREB (prolactin regulatory element binding protein). PA1-610 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~45 kDa protein representing recombit mouse PREB. The PA1-610 antigen is recombit mouse PREB.

Prolactin (PRL) expression in the pituitary is limited to specific cells. Pit-1 is a pituitary specific transcription factor that plays an important role in PRL expression, both in mature organism and during development. The PRL promoter contains numerous Pit-1 binding sites and these sites have been implicated in both basal level and kinase-mediated gene expression. The most proximal of these binding sites, termed 1P, has been shown to direct a response to numerous signals, such as cAMP and various G-proteins. A novel protein, termed PREB (prolactin regulatory element binding protein), has been recently identified that regulates PRL gene expression through the 1P site, though it contains no discernable DNA-binding motif. Recent studies suggest that PREB is encoded by a single-copy gene in both mice and humans and exhibits nuclear accumulation in pituitary cells. Evidence suggests that PREB is a novel transcription factor that assists in PRL expression whether alone, or in concert with Pit-1.
Specifications

Specifications

PREB
Polyclonal
PBS with 0.05% sodium azide
Q9WUQ2
Prolactin Regulatory Element Binding Protein
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
50907
Western Blot
Unconjugated
Preb
Liquid
PREB
Recombit mouse PREB.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit