Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pref-1/DLK1/FA1 Mouse anti-Human, Clone: 211309, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen Pref-1/DLK1/FA1
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pref-1 long isoform Ala24-Pro297 (with Arg248Pro and Lys295Ser substitutions) Accession # P80370
Clone 211309
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1144SP
View Documents
R&D Systems
MAB1144SP
25 ug
Each for $125.00
Add to cart
 
MAB1144100
View Documents
R&D Systems
MAB1144100
100 ug
Each for $377.00
Add to cart
 
MAB1144500
View Documents
R&D Systems
MAB1144500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Pref-1/DLK1/FA1 Monoclonal antibody MAB1144 Specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blotting
Specifications

Specifications

Pref-1/DLK1/FA1
211309
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
8788
RUO
Mouse myeloma cell line NS0-derived recombinant human Pref-1 long isoform Ala24-Pro297 (with Arg248Pro and Lys295Ser substitutions) Accession # P80370
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Purified
P80370
Monoclonal
Primary
Detects human Pref-1/DLK-1/FA1 in direct ELISAs and Western blots. Shows approximately 50% cross-reactivity with recombinant mouse Pref-1.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit