Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PREPL Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525478

Catalog No. PIPA525478


Add to cart

Description

Description

The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Specifications

Specifications

PREPL
Polyclonal
PBS with 0.09% sodium azide
Q4J6C6, Q8C167
putative prolyl oligopeptidase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100762687, 213760, 9581
Immunohistochemistry, Western Blot
Unconjugated
PREPL
Liquid
PREPL
KLH conjugated synthetic peptide between 624-652 amino acids from the C-terminal region of human PREPL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit