Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Presenilin 1 Chicken anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA11752

Catalog No. PA11752


Add to cart

Description

Description

PA1-1752 detects presenilin 1 protein (PS1) from human, rat and mouse samples. PA1-1752 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ~66 kDa protein representing PS1. The PA1-1752 immunogen is a synthetic peptides corresponding to residues K(311) N S K Y N A E S T E R(322) and E(342) A Q R D S H L G P H R(353) of human PS1.

Familial Alzheimer’s disease is often characterized by an early (<60 years of age) and rapid deterioration of the central nervous system. The symptoms are caused by the abnormal buildup of senile plaques composed of the 42 residue amyloid-beta peptide. This peptide is the result of the amyloid precursor protein being cleaved by the presenilin-gamma-secretase complex. Recent studies have linked mutations in presenilin 1 (PS1) and presenilin 2 (PS2) to a rapid increase in plaque accumulation. PS1 and PS2 are integral membrane proteins that contain 6-8 transmembrane domains and are predomitly localized within the endoplasmic reticulum and Golgi of neurons within the brain. Within the transmembrane regions of PS1 and PS2 there is over 60% homology, with the largest divergence found at the N-terminus and large loop region.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

Presenilin 1
Polyclonal
Unconjugated
PSEN1
Liquid
PSEN1
Mixture of synthetic peptides corresponding to residues K(311) N S K Y N A E S T E R(322) and E(342) A Q R D S H L G P H R(353) of human PS1.
PEG precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.02% sodium azide
P49768, P49769, P97887
PS1, PS-1, S182 protein
Chicken
IgY
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
19164, 29192, 5663
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit