Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSEN2 Chicken anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA11753

Catalog No. PA11753


Add to cart

Description

Description

PA1-1753 detects presenilin 2 protein (PS2) from human, rat and mouse samples. PA1-1753 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ~66 kDa protein representing PS2 The PA1-1753 immunogen is a mixture of synthetic peptides corresponding to residues Y(319) D P E N E E D S Y D S(330) and G(349) E E L E E E E E R G V(360) of human PS2.

Familial Alzheimer’s disease is often characterized by an early (<60 years of age) and rapid deterioration of the central nervous system. The symptoms are caused by the abnormal buildup of senile plaques composed of the 42 residue amyloid-beta peptide. This peptide is the result of the amyloid precursor protein being cleaved by the presenilin-gamma-secretase complex. Recent studies have linked mutations in presenilin 1 (PS1) and presenilin 2 (PS2) to a rapid increase in plaque accumulation. PS1 and PS2 are integral membrane proteins that contain 6-8 transmembrane domains and are predomitly localized within the endoplasmic reticulum and Golgi of neurons within the brain. Within the transmembrane regions of PS1 and PS2 there is over 60% homology, with the largest divergence found at the N-terminus and large loop region.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

PSEN2
Polyclonal
Unconjugated
PSEN2
Liquid
PSEN2
Mixture of synthetic peptides corresponding to residues Y(319) D P E N E E D S Y D S(330) and G(349) E E L E E E E E R G V(360) of human PS2.
PEG precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.02% sodium azide
P49810, Q61144, O88777
PS2, PS-2, STM-2, E5-1, AD3LP, AD5
Chicken
IgY
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
19165, 5664, 81751
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit