Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Presenilin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520376
Description
A suggested positive control is human brain tissue lysate. PA5-20376 can be used with blocking peptide PEP-0493.
Presenilin 1 Polyclonal antibody specifically detects Presenilin 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Presenilin 1 | |
Polyclonal | |
Unconjugated | |
PSEN1 | |
AD3; Ad3h; Alzheimer Disease 3; FAD; Minilin; presenilin 1; presenilin 1 (Alzheimer disease 3); presenilin1; presenilin-1; Presenilin-1 CTF subunit; Presenilin-1 CTF12; Presenilin-1 NTF subunit; Protein S182; PS1; PS-1; PS1-CTF12; PSEN1; Psnl1; S182; S182 protein; Senilin 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19164, 29192, 5663 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P49768, P49769, P97887 | |
PSEN1 | |
A 23 amino acid peptide from near the carboxy terminus of human presenilin1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction