Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha-1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525938

Catalog No. PIPA525938

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PRKAA1 is a protein belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5 ft.-prime-AMP-activated protein kinase. PRKAA1 is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of PRKAA1 is activated by the stimuli that increase the cellular AMP/ATP ratio. PRKAA1 regulates the activities of a number of key metabolic enzymes through phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways.
Specifications

Specifications

AMPK alpha-1
Polyclonal
PBS with 0.09% sodium azide
Q13131
AMPK, AMPKa1, 5′-AMP-activated protein kinase catalytic subunit alpha-1, 5′-AMP-activated protein kinase, catalytic alpha-1 chain, AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1, AMPK alpha 1, AMPK subunit alpha-1, tau-protein kinase P
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5562
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PRKAA1
Liquid
PRKAA1
recombit protein from human PRKAA1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit