Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha-2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525987
Description
This antibody is predicted to react with mouse, porcine and rat based on sequence homology.
The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.Specifications
AMPK alpha-2 | |
Polyclonal | |
PRKAA2 | |
AMPK, AMPK2, AMPKa2, PRKAA, 5′-AMP-activated protein kinase catalytic subunit alpha-2, 5′-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK subunit alpha-2, AMPK-alpha-2 chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5563 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
P54646 | |
PRKAA2 | |
KLH conjugated synthetic peptide between 145-173 amino acids from the central region of human PRKAA2 (Thr172) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction