Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKAR2A Mouse anti-Bovine, Human, Porcine, Clone: RS30, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA120070
Description
MA1-20071 detects PRKAR2 in human, bovine and porcine samples. MA1-20071 has been successfully used in ELISA and immunofluorescence procedures. The MA1-20071 immunogen is purified regulatory subunit of cAMP-dependent protein kinase type II from bovine b
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.Specifications
PRKAR2A | |
Monoclonal | |
Unconjugated | |
P00515, P13861, P05207 | |
PRKAR2A | |
Purified regulatory subunit of cAMP-dependent protein kinase type II from bovine brain. | |
100 μg | |
Primary | |
Bovine, Human, Porcine | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Immunofluorescence | |
RS30 | |
PRKAR2A | |
MGC3606, PKR2, PRKAR2 | |
Mouse | |
Protein G | |
RUO | |
100139910, 397493, 5576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title