Learn More
PKC delta Mouse anti-Human, Clone: 8E12, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519701
Catalog No. PIMA519701
Add to cart
Description
Description
Peptide Sequence: DILEKLFERE PTKRLGVTGN IKIHPFFKTI NWTLLEKRRL EPPFRPKVKS PRDYSNFDQE FLNEKARLSY SDKNLIDSMD QSAFAGFSFV NPKFEHLLED
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
Specifications
PKC delta
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Q05655
Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, Protein kinase C beta type, PKC-B, PKC-beta, PKCB, PRKCB1, PRKCB, Protein kinase C gamma type, PKC-gamma, PKCG, PRKCG, Protein kinase C delta type, Tyrosine-protein kinase PRKCD, nPKC-delta, Sphingosine-dependent protein kinase-1, SDK1, PRKCD, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2, PKC zeta, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5580
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
8E12
Unconjugated
PRKCD
Liquid
PRKCD
PRKCD (NP_006245, 577 a.a. ∽ 676 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human