Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein kinase, cGMP-dependent, type I, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PRKG1 protein.

Manufacturer:  Abnova Corporation H00005592B01

Catalog No. 89-006-030


Add to cart

Description

Description

Sequence: MGTLRDLQYALQEKIEELRQRDALIDELELELDQKDELIQKLQNELDKYRSVIRPATQQAQKQSASTLQGEPRTKRQAISAEPTAFDIQDLSHVTLPFYPKSPQSKDLIKEAILDNDFMKNLELSQIQEIVDCMYPVEYGKDSCIIKEGDVGSLVYVMEDGKVEVTKEGVKLCTMGPGKVFGELAILYNCTRTATVKTLVNVKLWAIDRQCFQTIMMRTGLIKHTEYMEFLKSVPTFQSLPEEILSKLADVLEETHYENGEYIIRQGARGDTFFIISKGTVNVTREDSPSEDPVFLRTLGKGDWFGEKALQG
Specifications

Specifications

protein kinase, cGMP-dependent, type I
Unconjugated
No additive
BC062688.1
CGKI|DKFZp686K042|FLJ36117|MGC71944|PGK|PKG|PRKG1B|PRKGR1B|cGKI-BETA|cGKI-alpha
Mouse
50 uL
Kinases
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human PRKG1 protein.
PRKG1
Serum
PRKG1
PRKG1 (AAH62688.1, 1 a.a. ~ 312 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5592
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit