Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130273

Catalog No. PIPA130273


Add to cart

Description

Description

PA1-30273 detects PRMT3 from human samples. PA1-30273 has been successfully used in Western blot applications. The PA1-30273 immunogen is: Synthetic peptide: STKTHWKQTVFLLEKPF, corresponding to amino acids 456-472 of Human PRMT 3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins (Tang et al., 1998 [PubMed 9642256]).
Specifications

Specifications

PRMT3
Polyclonal
Unconjugated
PRMT3
Liquid
PRMT3
synthetic peptide STKTHWKQTVFLLEKPF, corresponding to amino acids 456-472 of Human PRMT 3.
Protein G
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.6 mg/mL
PBS with 0.09% sodium azide
O60678
HRMT1L3
Rabbit
IgG
400 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10196
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit