Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Mouse anti-Human, Clone: 23C7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041460
Description
Use Anti-PRMT5 Antibody, clone 23C7 (mouse monoclonal antibody) validated in western blotting to detect PRMT5 also known as 72 kDa immunocytochemistryln-binding protein, HMT1 hnRNP methyltransferase-like 5, Jak-binding protein 1, SKB1 homolog.Specifications
PRMT5 | |
Monoclonal | |
Unconjugated | |
NP_001034708 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Western Blot | |
23C7 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
PRMT5, HRMT1L5, IBP72, SKB1Hs, JBP1, SKB1 | |
Recombinant protein corresponding to human PRMT5. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only