Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PRMT5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP136748

Catalog No. NBP136748

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PRMT5
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
NP_006100.2
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
Goat
IgG
0.1 mg
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
Primary
10419
Human
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
PRMT5
Peptide with sequence RACREKDRDPEAQ corresponding to internal region according to NP_001034708.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both isoforms (NP_006100.2; NP_001034708.1).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only