Learn More
Procainamide Mouse anti-Chemical, Clone: PC10-242.6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110763
Catalog No. 01-671-993
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10763 detects Procainamide. MA1-10763 has been successfully used in ELISA applications. The MA1-10763 immunogen is Procainamide, conjugated to KLH.
Procainamide is an oral antiarrhythmic agent that has been in wide use for more than 60 years. Long term procainamide therapy is known to induce hypersensitivity reactions, autoantibody formation and a lupus-like syndrome but is a rare cause of clinically apparent acute liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=4913]
Specifications
Specifications
Procainamide
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Procan; Procanbid; Biocoryl; Novocainamide; Novocamid; Pronestyl; Procaine amide; C13H21N3O
Procainamide, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
PC10-242.6
Unconjugated
Liquid
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical