Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procalcitonin Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA175361

Catalog No. PA175361

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-75361 specifically detects Procalcitonin. PA1-75361 has been successfully used in ELISA applications. The PA1-75361 immunogen is Recombit PCT.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Procalcitonin
Polyclonal
Unconjugated
CALCA
Liquid
CALCA
Recombit human PCT
Ig Fraction
RUO
Antibody
Polyclonal
Human
ELISA
9.31 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01258
PCT
Sheep
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
796
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit