Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRODH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527706

Catalog No. PIPA527706


Add to cart

Description

Description

PA5-27706 targets PRODH in WB applications and shows reactivity with Human samples. The PA5-27706 immunogen is recombit fragment corresponding to a region within amino acids 50 and 235 of Human PRODH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
Specifications

Specifications

PRODH
Polyclonal
Unconjugated
PRODH
Liquid
PRODH
Recombit fragment corresponding to a region within amino acids 50 and 235 of Human PRODH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O43272
HSPOX2, PIG6, POX, PRODH1, PRODH2, SCZD4, TP53I6, p53-induced gene 6 protein, proline dehydrogenase 1, mitochondrial, proline dehydrogenase, mitochondrial, proline oxidase 2, proline oxidase, mitochondrial, tumor protein p53 inducible protein 6
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5625
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit