Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Profilin 1, Polyclonal, Invitrogen™

Catalog No. PIPA131028
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA131028 100 μL
1 options

Catalog No. PIPA131028

Supplier: Thermo Scientific PA131028

Rabbit Polyclonal Antibody

The protein encoded by this gene is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals. Deletion of this gene is associated with Miller-Dieker syndrome.

PA1-31028 detects Profilin 1 from rat, Mouse, Marsupial, human, porcine samples.PA1-31028 has been successfully used in Western blot applications.The PA1-31028 immunogen is: Porcine and human platelet profilin affinity purified on the antigen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Profilin 1
Applications Western Blot
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Gene PFN1
Gene Accession No. P07737, P62962, P62963
Gene Symbols PFN1
Host Species Rabbit
Immunogen Porcine and human platelet profilin affinity purified on the antigen.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5216
Target Species Human, Marsupial, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.