Learn More
Progesterone Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175366
Catalog No. PA175366
Add to cart
Description
Description
PA1-75366 specifically detects Progesterone. PA1-75366 has been successfully used in ELISA applications. The PA1-75366 immunogen is Progesterone(4)-BTG.
Progesterone is the major progestational steroid, secreted primarily by the corpus luteum and the placenta. Progesterone binds to the progesterone receptor, resulting in dissociation of heat shock proteins, receptor phosphorylation, and transcription activation through direct or indirect interaction with transcription factors. Progesterone acts on the uterus, the mammary glands and the brain. It is required in embryo implantation, pregcy maintece, and the development of mammary tissue for milk production. Progesterone, converted from pregnenolone, also serves as an intermediate in the biosynthesis of gonadal steroid hormones and adrenal corticosteroids. Therapeutic progesterone is a synthetic form of the endogenous hormone progesterone. This agent exerts inhibitory effects on estrogens by decreasing the number of estrogen receptors and increasing its metabolism to inactive metabolites. [National Center for Biotechnology Information. PubChem Compound Database; CID=5994]
Specifications
Specifications
Progesterone
Polyclonal
PBS with 0.09% sodium azide
OHP; 11-alpha-hydroxyprogesterone; Hydroxyprogesterone alpha; hydroxyprogesterone 11; Progesterone-11a-hemisuccinate; Agolutin; Crinone; Luteohormone; Pregn-4-ene-3,20-dione; C21H30O2
Progesterone(4)-BTG
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical