Learn More
Progesterone Receptor Mouse anti-Human, Clone: 4D8, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA123268
Description
MA1-23268 detects Progesterone receptor in Human samples. MA1-23268 has been successfully used in Western Blot procedures. A suggested positive control for this product is MCF7. MA1-23268 immunogen corresponds to the region encoding amino acids 1-101 of the human progesterone receptor expressed in E. coli.. Store at 4°C short term. For extednded storage aliquot and store at -20°C or below, avoiding freeze/thaw cycles.
The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintece of pregcy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintece of the endometrium, maintece of pregcy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.Specifications
Progesterone Receptor | |
Monoclonal | |
1 mg/mL | |
PGR | |
PR, NR3C3, PGR | |
Mouse | |
Protein G | |
RUO | |
5241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
4D8 | |
Unconjugated | |
P06401 | |
PGR | |
The region encoding amino acids 1-101 of the human progesterone receptor expressed in E. coli. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.