Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Receptor, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138491

Catalog No. 01-673-876


Add to cart

Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities. Expression of PgR has been suggested to predict better clinical response to endocrine therapy than ER alone. PgR is phosphorylated on multiple serine residues (at least seven sites) through distinct groups of sites coordinately regulated by hormone and different kinases.

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is breast carcinoma.
Specifications

Specifications

Progesterone Receptor
Polyclonal
Assay Dependent
PGR
Liquid
PGR
Synthetic peptide derived from the C-terminus of human PgR
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P06401
PgR
Rabbit
IgG
1mL
4° C, do not freeze
Primary
5241
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit