Learn More
anti-Prolactin R, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232317
Catalog No. NBP232317
Add to cart
Description
Description
Prolactin R Polyclonal antibody specifically detects Prolactin R in Human samples. It is validated for Western Blot.
Specifications
Specifications
Prolactin R
0.5 mg/mL
Western Blot 1:1000
P16471, P16471
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Rabbit
IgG
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
PRLR
This PRLR antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 147-179 amino acids from the Central region of human PRLR.
69.506 kDa
400 ul
Apoptosis
Primary
5618