Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Prostaglandin E Synthase 2/PTGES2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133411

Catalog No. NBP133411


Add to cart

Description

Description

Prostaglandin E Synthase 2/PTGES2 Polyclonal antibody specifically detects Prostaglandin E Synthase 2/PTGES2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Prostaglandin E Synthase 2/PTGES2
0.44mg/mL
Western Blot 1:500-1:3000
Affinity Purified
PTGES2
Recombinant protein encompassing a sequence within the center region of human PTGES2. The exact sequence is proprietary.
42 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
C9orf15, chromosome 9 open reading frame 15, EC 5.3.99.3, FLJ14038, gamma-interferon-activated transcriptional element-binding factor 1, GATE-binding factor 1, GBF1, membrane-associated prostaglandin E synthase 2, Microsomal prostaglandin E synthase 2, microsomal prostaglandin E synthase-2, mPGES-2, PGES2MGC11289, prostaglandin E synthase 2
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
80142
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only