Learn More
anti-Prostatic Acid Phosphatase/ACPP, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP133600
Catalog No. NBP133600
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Prostatic Acid Phosphatase/ACPP
0.79mg/mL
Western Blot 1:500-1:3000
Affinity Purified
ACPP
Recombinant protein encompassing a sequence within the center region of human Prostatic Acid Phosphatase. The exact sequence is proprietary.
45 kDa
0.1 ml
Cell Cycle and Replication
Primary
55
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.