Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostein Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14230

Catalog No. PA14230


Add to cart

Description

Description

PA1-4230 detects the Prostein protein in human samples. PA1-4230 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~55 kDa protein representing Prostein in metastatic adenocarcinoma LNCaP.FGC cells. PA1-4230 immunogen is a mixture of synthetic peptides corresponding to the N- and C-terminal residues I(36) T Y V P P L L L E V G V E E(50) and F(537) A T Q V V F D K S D L A K Y S A(553) of human Prostein.

Prostein is a novel prostate-specific protein and marker expressed in normal and maligt prostate tissues. It is expressed in prostate tissues both at the plasma membrane and in the cytoplasm.
Specifications

Specifications

Prostein
Polyclonal
Unconjugated
SLC45A3
Liquid
SLC45A3
Synthetic peptides corresponding to residues I(36) T Y V P P L L L E V G V E E(50) and F(537) A T Q V V F D K S D L A K Y S A(553) of human Prostein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96JT2
Solute carrier family 45 member 3, Prostate cancer-associated protein 6
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
85414
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit