Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1971

Catalog No. PA1971

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum.The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman’s syndrome, and Liddle syndrome.

PA1-971 detects proteasome 19S subunit S10B from human cells.PA1-971 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects a 42kDa protein representing proteasome 19S subunit S10B from HeLa cell lysate. This antibody detects, to a lesser extent, an ~30kDa protein which could correspond to subunit degradation product.PA1-971 antigen is recombinant human proteasome 19S subunit S10B.
Specifications

Specifications

PSMD2
Polyclonal
PBS with 1mg/mL BSA and 0.05% sodium azide
Q13200
MGC14274, P97, Rpn1, S2, TRAP2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5708
Immunoprecipitation, Western Blot
Unconjugated
PSMD2
Liquid
PSMD2
Human recombit proteasome 19S subunit S10B.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit