Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1967

Catalog No. PA1967


Add to cart

Description

Description

PA1-967 detects proteasome 19S subunit S6 from human cells. PA1-967 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects a 48 kDa protein representing proteasome 19S subunit S6 from HeLa cell lysate. PA1-967 antigen is recombit human proteasome 19S subunit S6.

Proteolytic degradation is critical to the maintece of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman’s syndrome, and Liddle syndrome.
Specifications

Specifications

PSMC3
Polyclonal
PBS with 1mg/mL BSA and 0.05% sodium azide
P17980
MGC8487, TBP1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5702
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PSMC3
Liquid
PSMC3
Human recombit proteasome 19S subunit S6.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit