Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Proteasome 20S alpha 2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192292

Catalog No. NBP192292


Add to cart

Description

Description

PSMA2 Polyclonal antibody specifically detects PSMA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

Proteasome 20S alpha 2
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
EC 3.4.25.1, HC3PSC2, Macropain subunit C3, MU, Multicatalytic endopeptidase complex subunit C3, PMSA2, proteasome (prosome, macropain) subunit, alpha type, 2, Proteasome component C3, proteasome subunit alpha type-2, proteasome subunit HC3, PSC3
Rabbit
IgG
0.1 ml
Stem Cell Markers
Primary
5683
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
PSMA2
This antibody was developed against Recombinant Protein corresponding to amino acids:SGAYFAWKATAMGKNYVNGKTFLEKRYNEDLELEDAIHTAILTLKESFEGQMTEDNIEVGICNEAGFRRLTPTEVKDYLAAIA
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only