Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528999
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (91%), Dog (99%), Pig (97%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PSMB3 Polyclonal antibody specifically detects PSMB3 in Human samples. It is validated for Western Blot.Specifications
PSMB3 | |
Polyclonal | |
Unconjugated | |
PSMB3 | |
AL033320; C10-II; HC10-II; MGC4147; proteasome (prosome, macropain) subunit, beta type 3; proteasome (prosome, macropain) subunit, beta type, 3; proteasome beta 3 subunit; proteasome chain 13; Proteasome component C10-II; proteasome subunit beta 3; proteasome subunit beta type-3; Proteasome theta chain; PSMB3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.96 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P49720 | |
PSMB3 | |
Full length human Proteasome 20S beta 3 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction