Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB7 Rabbit anti-Bovine, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1979

Catalog No. PA1979

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit in the proteasome. Expression of this catalytic subunit is downregulated by gamma interferon and proteolytic processing is required to generate a mature subunit. This subunit is not present in the immunoproteasome and is replaced by catalytic subunit 2i (proteasome beta 10 subunit).

PA1-979 detects proteasome 20S Z (also known as the beta 2 subunit) from purified bovine and human 26S proteasome samples.PA1-979 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~29kDa protein representing proteasome 20S Z from purified human 26S proteasome.The PA1-979 immunizing peptides correspond to amino acid residues 263-277 from human proteasome 20S Z protein. This sequence is completely conserved in mouse and 97% conserved in rat protein. PA1-979 immunizing peptide (Cat. # PEP-175) is available for use in neutralization and control procedures.
Specifications

Specifications

PSMB7
Polyclonal
Unconjugated
PSMB7
Liquid
PSMB7
Synthetic Peptide: C E(263) I E V L E E T V Q T M D T S(NH2)(277)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q2TBP0, Q99436
RP11-101K10.7, Z
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
511207, 5695
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit