Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527521

Catalog No. PIPA527521


Add to cart

Description

Description

PA5-27521 targets Proteasome beta 4 in WB applications and shows reactivity with Human samples. The PA5-27521 immunogen is recombit fragment corresponding to a region within amino acids 1 and 264 of Human Proteasome beta 4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit.
Specifications

Specifications

PSMB4
Polyclonal
Unconjugated
PSMB4
Liquid
PSMB4
Recombit fragment corresponding to a region within amino acids 1 and 264 of Human Proteasome beta 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.73 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P28070
HN3, HsN3, PROS-26, PROS26, 26 kDa prosomal protein, hsBPROS26, macropain beta chain, multicatalytic endopeptidase complex beta chain, proteasome beta chain, proteasome chain 3, proteasome subunit HsN3, proteasome subunit beta type-4, proteasome subunit, beta type, 4
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5692
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit