Learn More
Protein A Goat anti-Bacteria, HRP, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA126853
Catalog No. PA126853
Add to cart
Description
Description
PA1-26853 detects Protein A. PA1-26853 has been successfully used in immunohistochemistry (frozen), ELISA, and Western blot procedures. The PA1-26853 immunogen is protein A from Staphylococcus aureus. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunglobulins.
Specifications
Specifications
Protein A
Polyclonal
HRP
Liquid
Protein A from Staphylococcus aureus.
IgG fraction
RUO
Antibody
Polyclonal
ELISA, Immunohistochemistry (Frozen), Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
Goat
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Bacteria