Learn More
Protein A Chicken anti-Bacteria, HRP, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Manufacturer:  Invitrogen PA172065
Catalog No. PA172065
Add to cart
Description
Description
PA1-72065 detects Protein A in bacteria samples. PA1-72065 has been successfully used in ELISA and western blot procedures. The PA1-72065 immunogen is from purified recombit protein A.
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunglobulins.
Specifications
Specifications
Protein A
Polyclonal
HRP
Liquid
Purified recombit protein A.
Protein A
RUO
Antibody
Polyclonal
ELISA, Western Blot
1.0 mg/mL
PBS with 50% glycerol and 0.01% thimerosal
Chicken
IgY
500 μg
-20°C
Primary
Bacteria