Learn More
Protein C Sheep anti-Mouse, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA143064
Catalog No. PA143064
Add to cart
Description
Description
PA1-43064 detects Protein C in mouse samples. PA1-43064 has successfully been used in Western blot and ELISA applications. PA1-43064 was derived from mouse protein C.
Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 µg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulmis, and recurrent venous thrombosis.
Specifications
Specifications
Protein C
Polyclonal
50%water with 50% glycerol and no preservative
P33587
PROC
Sheep
IgG
1 mg
-20° C, Avoid Freeze/Thaw Cycles
Primary
19123
ELISA, Western Blot
Unconjugated
Proc
Liquid
PROC
Murine Protein C.
Ammonium sulfate precipitation, Ion-exchange chromatography
RUO
Antibody
Polyclonal
Mouse