Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein capicua homolog (CIC) Mouse anti-Human, Rat, Clone: 6E12.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN449
Description
This Anti-Protein capicua homolog (CIC), clone 6E12.1 Antibody is validated for use in Immunohistochemistry (Paraffin) for the detection of Protein capicua homolog (CIC).
Specifications
Protein capicua homolog (CIC) | |
Monoclonal | |
1mg/mL | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
CIC | |
GST-tagged recombinant protein corresponding to human Protein capicua homolog (CIC). | |
100 μg | |
Human, Rat | |
Purified |
Immunohistochemistry (Paraffin) | |
6E12.1 | |
Unconjugated | |
NP_055940 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction