Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Gene Product 9.5 Rabbit anti-Human, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5925
Description
Detect Protein Gene Product 9.5 using this Anti-Protein Gene Product 9.5 Antibody validated for use in western blot, immunohistochemistry.Specifications
Protein Gene Product 9.5 | |
Polyclonal | |
Liquid with 0.05% sodium azide. | |
Rabbit | |
50 μL | |
NM_004181.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
UCHL1; PARK5; UCH-L1; Uch-L1; PGP9.5; EC 3.4.19.12; EC 6.-.-.- | |
Synthetic peptide corresponding to amino acids 108-127 of soluble cytoplasmic human PGP9.5. | |
Primary | |
Human, Mouse, Porcine, Rat | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction